Orders LinkContactUsLinkComponent
Dropping bottles with stoppers
Dropping bottles with stoppers
100 ml
Catalog # WITG5.883.100
Supplier:   witeg Labortechnik
Dropping bottles with stoppers
Catalog # WITG5.883.100
Supplier:  witeg Labortechnik
Dropping bottles with stoppers
Dropping bottles with stoppers
100 ml
Price
8,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
Glass, amber.
  • Good chemical resistance
  • Amber coloured to protect light sensitive substances
Dropping bottle with grooved flat stopper.
Specifications
  • Capacity:
    100 ml