Orders LinkContactUsLinkComponent
Spinner flasks, BIOGEN
Spinner flasks, BIOGEN
Supplier:   witeg Labortechnik
Spinner flasks, BIOGEN
Supplier:  witeg Labortechnik
Certificates
Spinner flasks, BIOGEN
9 Options Available Below
Product Details & Documents
Borosilicate glass with side arms
The fluid dynamic impeller assemblies to assure optimal lift of microcarrier beads at very low mixing speeds, the domed vessel bottom prevents microcarrier accumulations, positive-locking compression seal provides adjustable impeller assembly height, constructed of non-cytotoxic material, flasks inside surface siliconized on request.
Product Family Options
Product Information
  • Capacity
    Neck
    No. of side arms
    Ø×H
    Availability
    Price
  • 100 ml
    Centre neck 80 mm, side arm 18 mm
    2
    70×138 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 303,00 €
    303,00 €
    1 * 1 ST
       303,00 €
  • 250 ml
    Centre neck 80 mm, side arm 25 mm
    2
    90×176 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 328,00 €
    328,00 €
    1 * 1 ST
       328,00 €
  • 500 ml
    Centre neck 80 mm, side arm 32 mm
    2
    110×200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 337,00 €
    337,00 €
    1 * 1 ST
       337,00 €
  • 1000 ml
    Centre neck 110 mm, side arm 45 mm
    2
    138×246 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 446,00 €
    446,00 €
    1 * 1 ST
       446,00 €
  • 3000 ml
    Centre neck 110 mm, side arm 45 mm
    2
    186×358 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 623,00 €
    623,00 €
    1 * 1 ST
       623,00 €
  • 6000 ml
    Centre neck 110 mm, side arm 45 mm
    2
    200×420 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 791,00 €
    791,00 €
    1 * 1 ST
       791,00 €
  • 8000 ml
    Centre neck 110 mm, side arm 45 mm
    2
    224×443 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 814,00 €
    814,00 €
    1 * 1 ST
       814,00 €
  • 15000 ml
    Centre neck 110 mm, side arm 45 mm
    2
    300×570 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 939,00 €
    939,00 €
    1 * 1 ST
       939,00 €
  • 36000 ml
    Centre neck 110 mm, side arm 45 mm
    2
    400×600 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 360,00 €
    1 360,00 €
    1 * 1 ST
       1 360,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Screw cap, GL 100, PP, nature, with PTFE liner
    Screw cap, GL 100, PP, nature, with PTFE liner
    WITG0.175.099
    Log in for availability
    1 * 1 ST - 43,30 €