ContactUsLinkComponent
Cation exchange chromatography columns, CXpak series
Cation exchange chromatography columns, CXpak series
Supplier:  Resonac
Certificates
Cation exchange chromatography columns, CXpak series
Supplier:  Resonac
Specifications
  • Catalog No:
  • 554-2875
  • 554-2867
  • Description:
  • CXpak P-G
  • CXpak P-421S
  • Max. pressure:
  • 5,0 MPa (50 bar)
  • pH range:
  • ≥3
  • ≥3
  • Temperature limit:
  • 15 - 63 °C
  • 15 - 63 °C
  • USP code:
  • L22, L58
  • L22, L58