ContactUsLinkComponent
Cyanide distillation kit with Clear-Seal® joints, WHEATON®
Cyanide distillation kit with Clear-Seal® joints, WHEATON®
Catalog # WHEA377160
Supplier:  DWK Life Sciences
undefined
Cyanide distillation kit with Clear-Seal® joints, WHEATON®
Catalog # WHEA377160
Supplier:  DWK Life Sciences
Specifications
  • Description:
    Cyanide distillation kit complete
  • Pk:
    1
Specifications
About this item
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
  • The sample is reacted with H₂SO₄ and MgCl₂, which converts any cyanide to HCN
  • HCN distills over and is absorbed in a NaOH solution, which is then analyzed for CN
Ideal for removing interfering substances in quantitative analysis for cyanide.
Certifications: Standard Test Method for Cyanides in Water,# ASTM D2036