ContactUsLinkComponent
Reusable half masks, MX/PF
Reusable half masks, MX/PF
Catalog # 111-0548
Supplier:  HONEYWELL SAFETY
undefined
Reusable half masks, MX/PF
Catalog # 111-0548
Supplier:  HONEYWELL SAFETY
Specifications
  • Type:
    MX/PF F 950
  • Size:
    U*
  • Pk:
    1
Specifications
About this item
The half mask MX/PF is designed to work when the type of protection depends on the cartridges connected, gases, particles or combined. The APF is evaluated from 12 to 50.
  • Easy assembly
  • Silicone facepiece for additional comfort and easy decontamination
  • Triple flange seal
  • Wide, horizontal skin contact with pleated design
  • Good fit accross a wide variety of facial geometries
The half mask SPERIAN MX PF can be used only with certified T SERIES cartridges (following the standard EN 141 , EN 143)
Recommended for long term use