Orders LinkContactUsLinkComponent
Tree-pruning harness
Tree-pruning harness
1,78 kg
Catalog # DALL1002927
Supplier:   HONEYWELL SAFETY
Tree-pruning harness
Catalog # DALL1002927
Supplier:  HONEYWELL SAFETY
Tree-pruning harness
Tree-pruning harness
1,78 kg
Price
518,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Commonly used in conjunction with a climbing line, spikes and a work-positioning lanyard, these 2-point full body harnesses are suitable for heavier tree-pruning operations, for example when using a chainsaw. They feature a positioning belt, seat strap, rear anchorage for fall arrest and a sternal anchorage for the climbing line.
  • Climber is positioned against the trunk and is not required to move around the tree
  • Shoulder straps evenly distribute the weight of the tools (e.g. chainsaws) through the harness
  • Rigid seat strap provides a comfortable and safer position during work in suspension from the climbing line
  • Wide, padded belt is used in conjunction with the positioning lanyard around the tree-trunk
Specifications
  • Weight:
    1,78 kg