Filter tips
Supplier: BIOTIX
Certificates
About this item
This range of sterile filter pipette tips feature of the industry’s most efficient aerosol filter assemblies and reliably prevent aerosol transfer.
- Certified free of DNA, DNase, RNase and endotoxins
- Tips made from S3 polymer feature a sample saving surface for no hold-ups: Tips retain virtually no fluids, allowing users to dispense truly accurate sample volumes
- ESP™ reloading system allows reloading 10 trays in as little as 90 seconds: Saves time, lab space and the environment
Ideal for use when performing even the most sensitive assays.
Specifications
- Catalog No:
- 614-1424
- Environmentally Preferable:
- Safer Human and Environmental Health, Sustainable Packaging