Order Entry
Belgium
ContactUsLinkComponent
Ion chromatography columns, cation analysis
Ion chromatography columns, cation analysis
Supplier:  Resonac
Certificates
Ion chromatography columns, cation analysis
Supplier:  Resonac

Specifications

  • Catalog No:
  • 554-2896
  • 554-2935
  • 554-2887
  • 554-2936
  • Description:
  • IC YK-G
  • IC YK-421
  • IC YS-G
  • IC YS-50
  • Max. pressure:
  • 15 MPa (150 bar)
  • 15 MPa (150 bar)
  • pH range:
  • 2 - 7
  • 2 - 12
  • 2 - 12
  • Temperature limit:
  • 20 - 60 °C
  • 20 - 60 °C
  • 20 - 60 °C
  • 20 - 60 °C
  • USP code:
  • L76
  • L76
  • Storage:
  • 4 - 8 °C
  • 4 - 8 °C
  • 4 - 8 °C
  • 4 - 8 °C

Specifications

Line filters for ion chromatography

Product Information