ContactUsLinkComponent
Full face masks, Panoramasque
Full face masks, Panoramasque
Catalog # 111-1455
Supplier:  HONEYWELL SAFETY
undefined
Full face masks, Panoramasque
Catalog # 111-1455
Supplier:  HONEYWELL SAFETY
Specifications
  • Type:
    Silicone mask with RD filter connection and PMMA lens
  • Seq:
    04
  • Pk:
    1
Specifications
About this item
Panoramasque is a full face mask dedicated to offer respiratory protection, depending on the canister type assembled on it. For optimal protection, it is recommended to use aluminium or plastic Honeywell (Sperian) filters. They can only be used in sufficiently ventilated premises where the atmosphere contains a minimum oxygen volume of 17%.
  • Wide field of vision ensuring excellent lateral and downwards visibility
  • Skirt sealing flange for comfortable and tight seal
  • EPDM faceblank, making the mask lightweight and resistant
  • Good facial compatibility, one size with a high face
  • Five-strap head harness, quick-release adjusting buckles and fireproof cotton neck strap
  • Side lobes for wearing prescription spectacles inside mask
  • Lateral silicone exhalation valve for easy exhalation
  • Speech diaphragm ensures optimal communication
Certifications: EN 136:1989, APR; RD40 x 1/7" connection to mask conforming to the EN 148-1 standard