Order Entry
Belgium
ContactUsLinkComponent
SE 250 Mighty Small II Mini Vertical Electrophoresis Unit
SE 250 Mighty Small II Mini Vertical Electrophoresis Unit
Catalog # 732-1752
undefined
SE 250 Mighty Small II Mini Vertical Electrophoresis Unit
Catalog # 732-1752

Specifications

  • Model:
    SE 250
  • Description:
    Mini vertical electrophoresis system, complete unit with 0,75 mm, SE 250
  • Seq:
    02
  • Glass plate size (W×H):
    100×80 mm
  • Spacers:
    0,75 mm
  • Unit W×H×D:
    165×160×160 mm
  • Max. temperature:
    45 °C
  • Humidity:
    Up to 80%
  • Weight:
  • Pk:
    1

Specifications

About this item

The SE250 Mighty Small II mini vertical protein electrophoresis unit is designed for quick screening and rapid electrophoretic separation.

  • Maximum glass plate size 10×8 cm
  • Efficient active cooling ensures sharp bands
  • Quick and easy to assemble
  • Requires minimal buffer

Easily upgradable to the SE260 (with optional accessories) resulting in the additional capability of accommodating both 10×8 and 10×10,5 cm gels, enabling a wider variety of precast gel options.

Delivery: Complete system includes lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, 2× casting clamp assemblies, 4× cams, 10× rectangular glass plates (10×8 cm), 2× alumina plates, notched (10×8 cm), 4× spring clamps, 2× combs (10-well, 0,75 mm thickness), 4× T-spacers (0,75 mm thickness), 2× well locating decals, and gel seal. Basic unit includes same as above without the combs, spacers and caster, which can be ordered separately.