Orders LinkContactUsLinkComponent
VWR®, Universal pH Electrodes
VWR®, Universal pH Electrodes
Supplier:   VWR Collection
VWR®, Universal pH Electrodes
Supplier:  VWR Collection
Certificates
VWR®, Universal pH Electrodes
VWR®, Universal pH Electrodes
20 Options Available Below
Product Details & Documents
Laboratory and field electrodes for general applications. Supplied with a fixed cable or plug head, VWR® electrodes can be connected to most pH meters on the market.
  • Comfortable handling
  • Reliable, rapid readings
  • Gel or liquid electrolyte, glass or epoxy body shaft
  • Available with different connections: Plug head or fixed cable (1 metre) with either DIN or BNC type plug
All VWR® electrodes are checked at two points during the manufacturing process and then lot stamped to ensure quality. They are supplied with a soaker storage bottle and cap to make sure the pH-sensitive glass and reference junction remain in top condition, and are instantly ready for use.
Delivery information: Includes pH-electrode only.
Product Family Options
Product Information
  • Type
    Material
    Electrolyte
    Diaphr.
    Ø×L
    Connection
    Availability
    Price
  • 662-1382
    Single junction
    Single junction
    SJ 113
    Epoxy
    Gel
    Ceramic
    12×120 mm
    1 m cable with BNC connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,70 €
    94,70 €
    1 * 1 ST
       94,70 €
  • 662-1383
    Single junction
    Single junction
    SJ 114
    Epoxy
    Gel
    Ceramic
    12×120 mm
    1 m cable with DIN connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
  • 662-1384
    Single junction
    Single junction
    SJ 115
    Epoxy
    Gel
    Ceramic
    12×120 mm
    Plug head (S7)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 106,00 €
    106,00 €
    1 * 1 ST
       106,00 €
  • 662-1395
    Single junction
    Single junction
    SJ 223
    Glass
    KCl 3 mol/L
    Ceramic
    12×120 mm
    1 m cable with BNC connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 145,00 €
    145,00 €
    1 * 1 ST
       145,00 €
  • 662-1396
    Single junction
    Single junction
    SJ 224
    Glass
    KCl 3 mol/L
    Ceramic
    12×120 mm
    1 m cable with DIN connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 144,00 €
    144,00 €
    1 * 1 ST
       144,00 €
  • 662-1397
    Single junction
    Single junction
    SJ 225
    Glass
    KCl 3 mol/L
    Ceramic
    12×120 mm
    Plug head (S7)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 145,00 €
    145,00 €
    1 * 1 ST
       145,00 €
  • 662-1385
    Double junction
    Double junction
    DJ 113
    Epoxy
    Gel
    Ceramic
    12×120 mm
    1 m cable with BNC connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 144,00 €
    144,00 €
    1 * 1 ST
       144,00 €
  • 662-1386
    Double junction
    Double junction
    DJ 114
    Epoxy
    Gel
    Ceramic
    12×120 mm
    1 m cable with DIN connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 146,00 €
    146,00 €
    1 * 1 ST
       146,00 €
  • 662-1387
    Double junction
    Double junction
    DJ 115
    Epoxy
    Gel
    Ceramic
    12×120 mm
    Plug head (S7)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 138,00 €
    138,00 €
    1 * 1 ST
       138,00 €
  • 662-1391
    Single junction
    Single junction
    SM 123 (semi-micro)
    Glass
    Gel
    Ceramic
    12/6×120 mm
    1 m cable with BNC connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 207,00 €
    207,00 €
    1 * 1 ST
       207,00 €
  • 662-1393
    Single junction
    Single junction
    SM 125 (semi-micro)
    Glass
    Gel
    Ceramic
    12/6×120 mm
    Plug head (S7)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 207,00 €
    207,00 €
    1 * 1 ST
       207,00 €
  • 662-1402
    Single junction
    Single junction
    SM 223 (semi-micro)
    Glass
    KCl 3 mol/L
    Ceramic
    12/6×120 mm
    1 m cable with BNC connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €
  • 662-1403
    Single junction
    Single junction
    SM 224 (semi-micro)
    Glass
    KCl 3 mol/L
    Ceramic
    12/6×120 mm
    1 m cable with DIN connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 207,00 €
    207,00 €
    1 * 1 ST
       207,00 €
  • 662-1404
    Single junction
    Single junction
    SM 225 (semi-micro)
    Glass
    KCl 3 mol/L
    Ceramic
    12/6×120 mm
    Plug head (S7)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 195,00 €
    195,00 €
    1 * 1 ST
       195,00 €
  • 662-1405
    Single junction
    Single junction
    SP 223 (spear)
    Glass
    KCl 3 mol/L
    Ceramic
    12/6×120 mm
    1 m cable with BNC connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 222,00 €
    222,00 €
    1 * 1 ST
       222,00 €
  • 662-1406
    Single junction
    Single junction
    SP 224 (spear)
    Glass
    KCl 3 mol/L
    Ceramic
    12/6×120 mm
    1 m cable with DIN connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 209,00 €
    209,00 €
    1 * 1 ST
       209,00 €
  • 662-1388
    Single junction
    Single junction
    SF 113 (flat)
    Epoxy
    Gel
    Ceramic
    12×120 mm
    1 m cable with BNC connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 243,00 €
    243,00 €
    1 * 1 ST
       243,00 €
  • 662-1390
    Single junction
    Single junction
    SF 115 (flat)
    Epoxy
    Gel
    Ceramic
    12×120 mm
    Plug head (S7)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 271,00 €
    271,00 €
    1 * 1 ST
       271,00 €
  • 662-1398
    Single junction
    Single junction
    SF 223 (flat)
    Glass
    KCl 3 mol/L
    Ceramic
    12×120 mm
    1 m cable with BNC connector
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 253,00 €
    253,00 €
    1 * 1 ST
       253,00 €
  • 662-1401
    Single junction
    Single junction
    SF 225 (flat)
    Glass
    KCl 3 mol/L
    Ceramic
    12×120 mm
    Plug head (S7)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 243,00 €
    243,00 €
    1 * 1 ST
       243,00 €
Cables
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • DIN connector cable, coaxial, Coax54
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 67,40 €
    67,40 €
    1 * 1 ST
       67,40 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • Radiometer connector cable, coaxial, Coax56
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,70 €
    87,70 €
    1 * 1 ST
       87,70 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • BNC connector cable, coaxial, Coax53
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 54,90 €
    54,90 €
    1 * 1 ST
       54,90 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • Lemo (metrohm ) connector, 1 m cable, coaxial, Coax57
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,70 €
    87,70 €
    1 * 1 ST
       87,70 €