Order Entry
Belgium
ContactUsLinkComponent
HE 33 Mini Submarine Horizontal Electrophoresis Unit
HE 33 Mini Submarine Horizontal Electrophoresis Unit
Catalog # HOEFHE33-8-1.5
HE 33 Mini Submarine Horizontal Electrophoresis Unit
Catalog # HOEFHE33-8-1.5

Specifications

  • Model:
    HE33
  • Description:
    Mini submarine unit, complete with 1,5 mm thick, 8-well comb
  • Seq:
    01
  • Buffer volume:
  • Gel size W×L:
    70×100 mm
  • Max. power:
    15 W
  • Sample capacity:
    32 samples
  • Unit W×L×H:
    130×240×70 mm
  • Pk:
    1

Specifications

About this item

The HE33 mini horizontal electrophoresis unit is designed for very fast separations of DNA restriction fragments in agarose gels.

  • Run a 7×10 cm gel in as little as 5 minutes
  • UV transparent running tray allows the user to image the gel without risk of damage due to handling
  • Variety of comb sizes enables customisation of sample throughput to fit laboratory requirements
  • Adjustable comb height gives complete control over well depth

To achieve passive cooling, simply fill the clear moulded base with 50% ethylene glycol coolant solution, seal it, and place it in a freezer or ice bath until ready to use.

Accessories information: All combs require HE31-BK comb back, ordered separately.
Delivery: Complete system includes buffer chamber assembly, safety lid with high voltage leads, running tray, casting tray, 4× foam gaskets, bubble level, 1,5 mm thick 8-well comb, and comb back. Basic unit does not include combs or comb back, which can be ordered separately.

Combs

Product Information