ContactUsLinkComponent
Combs for Max Submarine Horizontal Electrophoresis Unit, He 99X
Combs for Max Submarine Horizontal Electrophoresis Unit, He 99X
Catalog # HOEFHE91-BK
Combs for Max Submarine Horizontal Electrophoresis Unit, He 99X
Catalog # HOEFHE91-BK
Specifications
  • Description:
    Comb back and 2 screws
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
All combs require HE-91-BK comb back, ordered separately.