Order Entry
Belgium
ContactUsLinkComponent
Indomethacin
Indomethacin
Catalog # PROOMM0584.00
CAS Number:  
Indomethacin
Catalog # PROOMM0584.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
ADR:6.1,I
 :357.80 g/mol
EINECS:200-186-5
UN:2811
CAS Number:53-86-1
Formula:C₁₉H₁₆ClNO₄
MDL Number:MFCD00057095