Organic reference standard, Benzo[b]fluoranthene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20565000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:205-99-2
- Description:Benzo[b]fluoranthene 10 µg/ml in cyclohexane
- Environmentally Preferable:
- Formula:C₂₀H₁₂
- Description:Benzo[b]fluoranthene