Order Entry
Belgium
ContactUsLinkComponent
Fluoranthene-[D10] 200 µg/ml in isooctane (98% D), CIL
Fluoranthene-[D10] 200 µg/ml in isooctane (98% D), CIL
Catalog # PROOCIL-DLM21401.2
CAS Number:  
Fluoranthene-[D10] 200 µg/ml in isooctane (98% D), CIL
Catalog # PROOCIL-DLM21401.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    93951-69-0
  • Description:
    Fluoranthene-[D10] 200 µg/ml in isooctane (98% D)
  • Formula:
    C₁₆D₁₀
  • Description:
    Fluoranthene-[D10]

Specifications

About this item