Order Entry
Belgium
ContactUsLinkComponent
Fluoranthene-[D10] 10 µg/ml in methanol, Dr Ehrenstorfer
Fluoranthene-[D10] 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL20795100ME
Supplier:  EHRENSTORFER
CAS Number:  
Fluoranthene-[D10] 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL20795100ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    93951-69-0
  • Description:
    Fluoranthene-[D10] 10 µg/ml in methanol
  • Environmentally Preferable:
  • Formula:
    C₁₆D₁₀
  • Description:
    Fluoranthene-[D10]

Specifications

About this item