

4-(Dimethylamino)cinnamaldéhyde réactif chromogène pour les indoles et les flavanols, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:6203-18-5
:
Formula:C₁₁H₁₃NO
MDL Number:MFCD00007002
:175.23 g/mol
Melting Pt:138,..140 °C (lit.)
:M-Clarity™ (Niveau de qualité) = MQ200
EINECS:228-267-0
:GHS07
:GHS09
Storage Temperature:Congélateur
Specification Test Results
- Appearance (colour):Yellow to very dark yellow and orange to very dark orange
- Appearance (form):Powder
- Purity (HPLC area %):≥97.5 %
- Solubility (colour):Yellow to very dark yellow to very dark red
- Solubility (turbidity):Clear to hazy
- Solubility (method):250 mg in 5 ml chloroform/ethanol (1:1)
- Proton nmr spectrum:Conforms to structure
- Recommended retest period:48 months
Specification Test Results
Frequently Bought Together