Order Entry
Belgium
ContactUsLinkComponent
Lithium cobaltite (Lithium cobalt (III) oxide)
Lithium cobaltite (Lithium cobalt (III) oxide)
Catalog # ICNA0522108580
Supplier:  MP Biomedicals
CAS Number:  
Lithium cobaltite (Lithium cobalt (III) oxide)
Catalog # ICNA0522108580
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :97.87 g/mol
EINECS:235-362-0
CAS Number:12190-79-3
Formula:CoLiO₂
MDL Number:MFCD00049786