Order Entry
Belgium
ContactUsLinkComponent
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Irritant
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Catalog # SOUT0205-01
CAS Number:  
Irritant
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Catalog # SOUT0205-01
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :433.6214 g/mol
EINECS:229-506-1
 :GHS07
Storage Temperature:Freezer
CAS Number:6578-06-9
Formula:C₁₅H₁₅BrClN₂O₄P
MDL Number:MFCD00040636