Order Entry
Belgium
ContactUsLinkComponent
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate) substrate ≥99%
Irritant
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate) substrate ≥99%
Catalog # PIER34040
CAS Number:  
Irritant
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate) substrate ≥99%
Catalog # PIER34040
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • VISUAL:
    White to off-white crystalline solid, free of foreign matter.
  • WATER CONTENT:
    (KF) ≤ 2%
  • MOLAR ABSORPTIVITY:
    ≥ 5700 at ~292 nm in 10mM Diethanolamine, pH 10.3 at 25 °C
  • PURITY:
    (HPLC) ≥ 99%
  • INORGANIC PHOSPHATE:
    ≤ 0.1%

Specification Test Results

About this item

 :
 :433.6214 g/mol
EINECS:229-506-1
 :GHS07
Storage Temperature:Freezer
CAS Number:6578-06-9
Formula:C₁₅H₁₅BrClN₂O₄P
MDL Number:MFCD00040636