Digoxin
Glass bottle for solids, 50 mg
Catalog # ROTH7323.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:20830-75-5
- Formula:C₄₁H₆₄O₁₄
- MDL Number:MFCD00003674
- Molecular Weight:780.95 g/mol
- EINECS:244-068-1
- UN:2811
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg



