Order Entry
Belgium
Orders LinkContactUsLinkComponent
Digoxin

Digoxin

Glass bottle for solids, 100 mg
Catalog # PROOICRS0289
Digoxin
Catalog # PROOICRS0289
CAS Number:  20830-75-5
Digoxin

Glass bottle for solids, 100 mg

CAS Number:  20830-75-5
Price
367,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
  • CAS Number:20830-75-5
  • Formula:C₄₁H₆₄O₁₄
  • MDL Number:MFCD00003674
  • Molecular Weight:780.95 g/mol
  • EINECS:244-068-1
  • UN:2811
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.