Order Entry
Belgium
Orders LinkContactUsLinkComponent
Digoxin

Digoxin

Glass bottle for solids, 10 mg
Catalog # EDQMY0000554
Supplier:   EDQM
Digoxin
Catalog # EDQMY0000554
Supplier:  EDQM
CAS Number:  20830-75-5
Digoxin

Glass bottle for solids, 10 mg

CAS Number:  20830-75-5
Price
270,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
  • CAS Number:20830-75-5
  • Formula:C₄₁H₆₄O₁₄
  • MDL Number:MFCD00003674
  • Molecular Weight:780.95 g/mol
  • EINECS:244-068-1
  • UN:2811
  • ADR:6.1,II

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.