Digoxin 96%
Fournisseur: Thermo Fisher Scientific
Jump to:
About this item
Purity: 96% CAS No.: 20830-75-5 Molecular Formula: C41H64O14 Formula Weight: 780.95 Melting Point: ca 250° dec. Merck Reference: 14,3167 MDL No.: MFCD00003674
- CAS Number:20830-75-5
- Formula:C₄₁H₆₄O₁₄
- MDL Number:MFCD00003674
- Molecular Weight:780.95 g/mol
- Merck Index:14,3167
- Melting Pt:∼ 250 °C (dec.)
- Storage Temperature:Ambient
- EINECS:244-068-1
- UN:2811
- ADR:6.1,II
3 Options Available Below
Product Details & Documents
Purity: 96% CAS No.: 20830-75-5 Molecular Formula: C41H64O14 Formula Weight: 780.95 Melting Point: ca 250° dec. Merck Reference: 14,3167 MDL No.: MFCD00003674
- CAS Number:20830-75-5
- Formula:C₄₁H₆₄O₁₄
- MDL Number:MFCD00003674
- Molecular Weight:780.95 g/mol
- Merck Index:14,3167
- Melting Pt:∼ 250 °C (dec.)
- Storage Temperature:Ambient
- EINECS:244-068-1
- UN:2811
- ADR:6.1,II
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



