Tetrakis(decyl)ammonium bromide, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Tetrakis(decyl)ammonium bromide, IPC-TDA-Br, Tetra(decyl)ammonium bromide
CAS Number:14937-42-9
:
Formula:C₄₀H₈₄BrN
MDL Number:MFCD00043166
Molecular Weight:659.0057 g/mol
:M-Clarity™ quality level = MQ100
EINECS:239-013-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together