Order Entry
Belgium
ContactUsLinkComponent
Lead(II) nitrate 0.1 mol/l (0.2 N) in aqueous solution
Health
Environment
Lead(II) nitrate 0.1 mol/l (0.2 N) in aqueous solution
Catalog # 307227.0500
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Environment
Lead(II) nitrate 0.1 mol/l (0.2 N) in aqueous solution
Catalog # 307227.0500
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :331.21 g/mol
EINECS:233-245-9
UN:3082
ADR:9,III
 :GHS08
 :GHS09
CAS Number:10099-74-8
Formula:Pb(NO₃)₂
MDL Number:MFCD00011153