Order Entry
Belgium
ContactUsLinkComponent
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Flammable
Toxic
Corrosive
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Catalog # 84574.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Toxic
Corrosive
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Catalog # 84574.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.5 %
  • Appearance:
    Clear liquid
  • Colour value:
    Max. 25.0 APHA
  • Identity (IR):
    Passes test
  • Primary and secondary amines:
    Max. 0.002 %
  • Residue on evaporation:
    Max. 0.001 %
  • Water:
    Max. 0.03 %

Specification Test Results

About this item

 :
 :129.25 g/mol
Boiling Pt:127 °C (1013 hPa)
Melting Pt:–50 °C
Density:0.76 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:230-392-0
UN:2733
ADR:3 (8),II
 :GHS02
 :GHS06
 :GHS05
CAS Number:7087-68-5
Formula:C₈H₁₉N
MDL Number:MFCD00008868