Order Entry
Belgium
Orders LinkContactUsLinkComponent
Test tubes without rim

Test tubes without rim

20 ml, 150 mm, 16 mm
Catalog # LPIT110118
Supplier:   LP ITALIANA
Test tubes without rim
Catalog # LPIT110118
Supplier:  LP ITALIANA
Test tubes without rim
Test tubes without rim

20 ml, 150 mm, 16 mm

Price
420,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

PS, clear.

  • Optimal visibility of contents
  • Good chemical resistance
  • Good temperature resistance

Universal tubes with flat, round or conical bottom for a wide range of applications.

Specifications
  • Ext.Ø:
    16 mm
  • Capacity:
    20 ml
  • Length:
    150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.