Microcentrifuge tubes, 1,5 ml
LPIT111540
- Type:with attached snap cap, Eppendorf type
- Version:Graduated, conical bottom, non sterile
- Capacity:1,5 ml
- Colour:Natural
- Environmentally Preferable:
- Pk:10.000
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
- Graduated.
With flat cap.