Rotors for ROTOFIX 46/46H centrifuges
Catalog # 521-1232
Fournisseur: Hettich
Spécifications
- Description:Swing-out rotor, 4-place, for ASTM tubes, angle 90°, 2000 min⁻¹, 984×g
- Seq:01
- Environmentally Preferable:Low Manufacturing Impact
- Pk:1
- For:ROTOFIX 46/46H/ROTANTA
- Environmental certification:ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)