Orders LinkContactUsLinkComponent
VWR® Separating Funnels, Squibb
VWR® Separating Funnels, Squibb
Supplier:  VWR Collection
Certificates
VWR® Separating Funnels, Squibb
VWR® Separating Funnels, Squibb
12 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance
Product Family Options
Without graduation, with PTFE key
Product Information
With graduation, with PTFE key
Product Information