Orders LinkContactUsLinkComponent
VWR®, Separating Funnels, Conical
VWR®, Separating Funnels, Conical
VWR®, Separating Funnels, Conical
VWR®, Separating Funnels, Conical
Borosilicate 3.3 glass, with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance
Product Family Options
With PTFE key, non graduated
  • Capacity
    Socket size
  • 50 ml
    19/26 NS
    1 * 1 ST
    1 * 1 ST - 49,20 €
    49,20 €
    1 * 1 ST
       49,20 €
  • 100 ml
    19/26 NS
    1 * 1 ST
    1 * 1 ST - 50,00 €
    50,00 €
    1 * 1 ST
       50,00 €
  • 250 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 63,30 €
    63,30 €
    1 * 1 ST
       63,30 €
  • 500 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 72,10 €
    72,10 €
    1 * 1 ST
       72,10 €
  • 1000 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 96,20 €
    96,20 €
    1 * 1 ST
       96,20 €
  • 2000 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 148,00 €
    148,00 €
    1 * 1 ST
       148,00 €
With PTFE key, graduated
  • Capacity
    Socket size
  • 50 ml
    19/26 NS
    1 * 1 ST
    1 * 1 ST - 52,10 €
    52,10 €
    1 * 1 ST
       52,10 €
  • 100 ml
    19/26 NS
    1 * 1 ST
    1 * 1 ST - 53,30 €
    53,30 €
    1 * 1 ST
       53,30 €
  • 250 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 65,90 €
    65,90 €
    1 * 1 ST
       65,90 €
  • 500 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 77,40 €
    77,40 €
    1 * 1 ST
       77,40 €
  • 1000 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 101,00 €
    101,00 €
    1 * 1 ST
       101,00 €
  • 2000 ml
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 157,00 €
    157,00 €
    1 * 1 ST
       157,00 €