Orders LinkContactUsLinkComponent
Dissection boards, disposable
Dissection boards, disposable
Supplier:   LP ITALIANA
Dissection boards, disposable
Supplier:  LP ITALIANA
Certificates
Dissection boards, disposable
Dissection boards, disposable
2 Options Available Below
Product Details & Documents
Disposable cutting boards made of cardboard and PS foam for the dissection of surgical specimens.
  • With data registration area
  • Pre-printed scale (in mm)
  • Three different sizes available
Product Family Options
Product Information
  • Colour
    L×W
    Availability
    Price
  • LPIT194032
    Disposable dissection boards
    Disposable dissection boards
    White
    333×233 mm
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 457,00 €
    457,00 €
    1 * 48 ST
       457,00 €
  • LPIT197032
    Disposable dissection boards
    Disposable dissection boards
    White
    350×333 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 322,00 €
    322,00 €
    1 * 24 ST
       322,00 €