Orders LinkContactUsLinkComponent
Wire baskets, square
Wire baskets, square
Wire baskets, square
Certificates
Wire baskets, square
Wire baskets, square
2 Options Available Below
Product Details & Documents
These square baskets are made of stainless steel. Designed for transporting, storing, cleaning and sterilising laboratory utensils.
  • Excellent temperature resistance
  • Good chemical resistance
  • Reinforced edges
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 160×160×160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 143,00 €
    143,00 €
    1 * 1 ST
       143,00 €
  • 200×200×200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 177,00 €
    177,00 €
    1 * 1 ST
       177,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.