VWR®, Incinerating Dishes
Supplier: VWR Collection
Certificates
About this item
Porcelain, glazed, produced according to DIN 12904.
- Dishes can withstand temperatures up to 1000 °C
- Stable with large flat unglazed base
- Glazed porcelain has high resistance to acids and bases
Frequently Bought Together