Orders LinkContactUsLinkComponent
RevertAid™ Reverse Transcriptase
RevertAid™ Reverse Transcriptase
RevertAid™ Reverse Transcriptase
Certificates
RevertAid™ Reverse Transcriptase
RevertAid™ Reverse Transcriptase
2 Options Available Below
Product Details & Documents
RevertAid™ Reverse Transcriptase (RT) is a genetically modified M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity and a RNase H activity specific to RNA in RNA-DNA hybrids, which is significantly lower than that of Avian Myeloblastosis Virus (AMV) reverse transcriptase.
  • Efficient synthesis of full length first strand cDNA up to 13 kb
  • Optimum activity at 42 °C
  • Active up to 50 °C
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labelled nucleotides)
Delivery information: Supplied with 5X reaction buffer.
Documents
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • EP0441
    RevertAid™ Reverse Transcriptase, 10,000 units (200 U/µl), for 50 reactions of 20 µl
    RevertAid™ Reverse Transcriptase, 10,000 units (200 U/µl), for 50 reactions of 20 µl
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 98,30 €
    98,30 €
    1 * 1 KIT
       98,30 €
  • EP0442
    RevertAid™ Reverse Transcriptase, 5×10,000 units (200 U/µl), for 250 reactions of 20 µl
    RevertAid™ Reverse Transcriptase, 5×10,000 units (200 U/µl), for 250 reactions of 20 µl
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 373,00 €
    373,00 €
    1 * 1 KIT
       373,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.