Orders LinkContactUsLinkComponent
Receiving flasks
Receiving flasks
Supplier:   BUCHI
Receiving flasks
Supplier:  BUCHI
Certificates
Receiving flasks
Receiving flasks
12 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for BÜCHI rotary evaporators. With spherical joint socket KS 35/20.
Product Family Options
Product Information
  • Capacity
    Type
    Socket
    Availability
    Price
  • 1000 ml
    uncoated
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 126,00 €
    126,00 €
    1 * 1 ST
       126,00 €
  • 2000 ml
    uncoated
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 244,00 €
    244,00 €
    1 * 1 ST
       244,00 €
  • 3000 ml
    uncoated
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 287,00 €
    287,00 €
    1 * 1 ST
       287,00 €
  • 2000 ml
    P+G coated
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 284,00 €
    284,00 €
    1 * 1 ST
       284,00 €
  • 50 ml
    uncoated
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
  • 100 ml
    uncoated
    KS35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 114,00 €
    114,00 €
    1 * 1 ST
       114,00 €
  • 250 ml
    uncoated
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • 500 ml
    uncoated
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 121,00 €
    121,00 €
    1 * 1 ST
       121,00 €
  • 500 ml
    Low temperature
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 182,00 €
    182,00 €
    1 * 1 ST
       182,00 €
  • 2000 ml
    Low temperature
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 278,00 €
    278,00 €
    1 * 1 ST
       278,00 €
  • 1000 ml
    Low temperature
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 182,00 €
    182,00 €
    1 * 1 ST
       182,00 €
  • 3000 ml
    Low temperature
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 349,00 €
    349,00 €
    1 * 1 ST
       349,00 €