Ladles
Supplier: Sampling Systems
Jump to:
About this item
These ladles made from hygienic stainless steel are ideal for sampling liquid products. The ladles have a crevice free construction so that they can be easily cleaned between batches.
- Crevice free construction allows easy cleaning between batches
- Satin finish
5 Options Available Below
Product Details & Documents
These ladles made from hygienic stainless steel are ideal for sampling liquid products. The ladles have a crevice free construction so that they can be easily cleaned between batches.
- Crevice free construction allows easy cleaning between batches
- Satin finish
Product Family Options
Product Information
- CapacityLengthØMaterialAvailabilityPrice