Orders LinkContactUsLinkComponent
SteriWare® Liquid Funnels
SteriWare® Liquid Funnels
SteriWare® Liquid Funnels
Supplier:  Sampling Systems
Certificates
SteriWare® Liquid Funnels
SteriWare® Liquid Funnels
8 Options Available Below
Product Details & Documents
Single use, disposable liquid funnels ideal for laboratories and manufacturing areas. Designed to use straight from bags.
  • Made of HDPE
  • Moulded and packed in a class 100,000 cleanroom
  • With small hygenic side handle
  • BSE/TSE free
  • Available sterile (gamma irradiated)
Certifications: Material conforms to FDA, EU 10/2011 and EC 1935/2004. Full batch traceability.
Packaging: Individually bagged.
Product Family Options
Product Information
  • Height
    Length
    Stem O-Ø
    Int.Ø top
    Availability
    Price
  • 300-0590
    Funnel, natural
    Funnel, natural
    8 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 186,00 €
    186,00 €
    1 * 100 ST
       186,00 €
  • 300-0591
    Funnel, natural, sterile
    Funnel, natural, sterile
    8 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 252,00 €
    252,00 €
    1 * 100 ST
       252,00 €
  • 300-0589
    Funnel, blue
    Funnel, blue
    10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 93,80 €
    93,80 €
    1 * 50 ST
       93,80 €
  • SAMP8370B-100S
    Funnel, blue, sterile
    Funnel, blue, sterile
    10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 224,00 €
    224,00 €
    1 * 50 ST
       224,00 €
  • 300-0592
    Funnel, natural
    Funnel, natural
    11 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 91,40 €
    91,40 €
    1 * 50 ST
       91,40 €
  • Sterile
    103 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 210,00 €
    210,00 €
    1 * 50 ST
       210,00 €
  • SAMP8370H-200
    Funnel, natural
    Funnel, natural
    19 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 64,20 €
    64,20 €
    1 * 10 ST
       64,20 €
  • SAMP8370H-200S
    Funnel, natural, sterile
    Funnel, natural, sterile
    180 mm
    19 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 188,00 €
    188,00 €
    1 * 10 ST
       188,00 €