Orders LinkContactUsLinkComponent
Erlenmeyer flasks with screw caps
Erlenmeyer flasks with screw caps
Supplier:   witeg Labortechnik
Erlenmeyer flasks with screw caps
Supplier:  witeg Labortechnik
Certificates
Erlenmeyer flasks with screw caps
Erlenmeyer flasks with screw caps
5 Options Available Below
Product Details & Documents
Erlenmeyer flasks, made of borosilicate glass, with screw-thread, graduated can be used for the storage of culture media and mixing.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Thread
    Availability
    Price
  • 100 ml
    105 mm
    64 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,40 €
    19,40 €
    1 * 1 ST
       19,40 €
  • 1000 ml
    220 mm
    131 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,40 €
    28,40 €
    1 * 1 ST
       28,40 €
  • 250 ml
    140 mm
    85 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,90 €
    22,90 €
    1 * 1 ST
       22,90 €
  • 500 ml
    175 mm
    105 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,30 €
    24,30 €
    1 * 1 ST
       24,30 €
  • 2000 ml
    280 mm
    166 mm
    GL 45
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,20 €
    59,20 €
    1 * 1 ST
       59,20 €