Pasteur pipettes
Supplier: LP ITALIANA
Jump to:
About this item
Disposable Pasteur pipettes made from plastic.
7 Options Available Below
Product Details & Documents
Disposable Pasteur pipettes made from plastic.
Product Family Options
Product Information
- CapacityLengthGraduationGraduated up toSterilePackedAvailabilityPrice
- LPIT135038Pasteur pipette, graduatedPasteur pipette, graduated7,0 ml150 mm0,5 - 3 ml3 ml+In bags of 20Log in for availability1 * 1.000 ST186,00 €
- LPIT135138Pasteur pipette, graduatedPasteur pipette, graduated7,0 ml150 mm0,5 - 3 ml3 ml+Individually wrappedLog in for availability1 * 1.000 ST234,00 €
- LPIT137030Pasteur pipette, graduatedPasteur pipette, graduated5,0 ml150 mm0,25 - 1 ml1 ml-In box of 500Log in for availability1 * 5.000 ST430,00 €
- 613-4502Pasteur pipette, graduatedPasteur pipette, graduated5,0 ml150 mm0,25 - 1 ml1 ml+In bags of 20Log in for availability1 * 1.000 ST182,00 €
- LPIT137138Pasteur pipette, graduatedPasteur pipette, graduated5,0 ml150 mm0,25 - 1 ml1 ml+Individually wrappedLog in for availability1 * 1.000 ST229,00 €
- 613-4504Pasteur pipette, graduatedPasteur pipette, graduated6,0 ml230 mm0,75 - 1,5 ml1,5 ml+In bags of 20Log in for availability1 * 1.000 ST268,00 €
- LPIT139138Pasteur pipette, graduatedPasteur pipette, graduated6,0 ml230 mm0,75 - 1,5 ml1,5 ml+Individually wrappedLog in for availability1 * 1.000 ST553,00 €