Orders LinkContactUsLinkComponent
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
Supplier:   witeg Labortechnik
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
Supplier:  witeg Labortechnik
Certificates
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
Multi-channel pipettes, mechanical, variable volume, WITOPET-Multi
6 Options Available Below
Product Details & Documents
These multi-channel pipettes are designed for comfort over long term repetitive pipetting. 8 or 12 identical volumes for titration plates.
  • Highest possible accuracy and precision
  • Easy readable digital volume display
  • Volume ranges colour coded on the push button
  • Volumes steplessly adjustable
  • Tip ejector can be blocked to avoid mis-application when using the instrument over large series
Delivery information: Each pipette is individually tested and delivered with a test certificate.
Product Family Options
8-channel
Product Information
  • Volume
    Increment
    Accuracy
    Imprecision
    Availability
    Price
  • ME 10
    0,5 - 10 µl
    0,1 µl
    ≤ ±1,5 - ±4,0 %
    ≤ ±1,5 - ±4,0 %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 404,00 €
    404,00 €
    1 * 1 ST
       404,00 €
  • ME 50
    5 - 50
    1,0 µl
    ≤ ±1,0 - ±3,0 %
    ≤ ±0,5 - ±2,0 %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 404,00 €
    404,00 €
    1 * 1 ST
       404,00 €
  • ME 300
    50 - 300 µl
    1,0 µl
    ≤ ±0,7 - ±1,5 %
    ≤ ±0,25 - ±0,8 %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 404,00 €
    404,00 €
    1 * 1 ST
       404,00 €
12-channel
Product Information
  • Volume
    Increment
    Accuracy
    Imprecision
    Availability
    Price
  • ME 10
    0,5 - 10 µl
    0,1 µl
    ≤ ±1,5 - ±4,0 %
    ≤ ±1,5 - ±4,0 %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • ME 50
    5 - 50 µl
    1,0 µl
    ≤ ±1,0 - ±3,0 %
    ≤ ±0,5 - ±2,0 %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • ME 300
    50 - 300 µl
    1,0 µl
    ≤ ±0,7 - ±1,5 %
    ≤ ±0,25 - ±0,8 %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €