Order Entry
Belgium
ContactUsLinkComponent
Test tube racks, PP
Test tube racks, PP
Catalog # LPIT180042
Supplier:  LP ITALIANA
undefined
Test tube racks, PP
Catalog # LPIT180042
Supplier:  LP ITALIANA

Specifications

  • For tube Ø:
    30 mm
  • No. of Holes:
    18 (3×6)
  • Colour:
    White
  • W×D×H:
    243×126×64 mm
  • Pk:
    15

Specifications

About this item

Racks, made of Polypropylene, white.

  • Stable
  • Excellent chemical resistance
  • Excellent temperature resistance