Volumetric flasks, trapezoidal shape, class A, brown or blue markings
Supplier: witeg Labortechnik
Certificates
About this item
Borosilicate glass 3.3 with PE stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
- Brown or blue markings
Certifications: DIN EN ISO 1042
Frequently Bought Together