ContactUsLinkComponent
Volumetric flasks, class A, brown markings
Volumetric flasks, class A, brown markings
Catalog # 613-4125
Supplier:  witeg Labortechnik
undefined
Volumetric flasks, class A, brown markings
Catalog # 613-4125
Supplier:  witeg Labortechnik
Specifications
  • NS:
    10/19
  • Capacity:
    15 ml
  • Pk:
    2
  • Tolerance:
    0,04 ± ml
Specifications
About this item
DURAN® borosilicate glass 3.3 with PE stopper.
  • With spout
  • Conformity certified
  • Delivered with batch number and have enclosed one batch certificate per package