Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue stainer, MSM/24

Tissue stainer, MSM/24

Tissue stainer, MSM/24, 30
Catalog # SLEE12000205
Supplier:   Slee
Tissue stainer, MSM/24
Catalog # SLEE12000205
Supplier:  Slee
Tissue stainer, MSM/24
Tissue stainer, MSM/24

Tissue stainer, MSM/24, 30

Price
18 750,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The MSM/24 carousel stainer with increased user safety with fume extraction efficiently eliminates hazardous fumes.

  • Integrated battery ensuring highest process
  • Safety even in environments with frequent power failures

Designed to meet the requirements for routine staining in histology and cytology laboratories.

Delivery information: Supplied with operation manual, 24 plastic cuvettes, transport basket (plastic) with 30 standard size micro slides, and main cable.

Specifications
  • Model:
    MSM/24
  • No. of slides:
    30
  • Description:
    Tissue stainer
  • Cat. No.:
    SLEE12000205
  • No. of programs:
    20
  • W×D×H:
    870×870×700 mm
  • Electrical:
    100 − 240 V/50 − 60 Hz
  • Weight:
    80 kg
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Exhaust fan with carbon filter
    Exhaust fan with carbon filter
    SLEE11000211
    Log in for availability
    1 * 1 ST - 1 000,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Perspex hood
    Perspex hood
    SLEE11000220
    Log in for availability
    1 * 1 ST - 1 640,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Drying station
    Drying station
    SLEE12000256
    Log in for availability
    1 * 1 ST - 1 150,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.