Orders LinkContactUsLinkComponent
Magnetic stirrers, boxed sets
 :   COWIE
Magnetic stirrers, boxed sets
 :  COWIE
Magnetic stirrers, boxed sets
Magnetic stirrers, boxed sets
Boxed sets include an assortment of useful sizes of each type of PTFE stirrer bar in a re-usable container.
Product Family Options
  • Type
  • Plain, 2 each of 10×6, 15×6, 20×7, 25×8, 30×7, 40×8, 50×8, 60×7 and 80×10 mm
    1 * 1 SET
    1 * 1 SET - 52,70 €
    52,70 €
    1 * 1 SET
       52,70 €
  • Tapered, 2 each of 10×4, 15×5, 20×7, 25×8, 30×8, 35×8, 40×8, 50×8, 60×8 mm, 1 each of 70×10, 80×10 mm and 1 each of 300×10 mm retriever
    1 * 1 ST
    1 * 1 ST - 121,00 €
    121,00 €
    1 * 1 ST
       121,00 €
  • Turbo elliptical, 3 each of 10×6, 15×10, 25×14 mm and 1 each of 50×24 mm
    1 * 1 ST
    1 * 1 ST - 64,40 €
    64,40 €
    1 * 1 ST
       64,40 €
  • Turbo cylindrical, 2 each of 8×3, 12×4,5, 20×6, 25×6, 30×6, 40×8 mm and 1 each of 50×8 and 60×10 mm
    1 * 1 ST
    1 * 1 ST - 75,70 €
    75,70 €
    1 * 1 ST
       75,70 €