Orders LinkContactUsLinkComponent
Separating funnels, cylindrical
Separating funnels, cylindrical
100 ml, 19/26 NS
Catalog # WITG2.643.100
Supplier:   witeg Labortechnik
Separating funnels, cylindrical
Catalog # WITG2.643.100
Supplier:  witeg Labortechnik
Separating funnels, cylindrical
Separating funnels, cylindrical
100 ml, 19/26 NS
Price
59,80 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Cylindrical separatory funnels, made of borosilicat glass, with Witaflo PTFE needle valve stopcock.
  • Graduated
  • Very good chemical resistance
  • Very good temperature resistance
Spécifications
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml