Orders LinkContactUsLinkComponent
Separating funnels, spherical
Separating funnels, spherical
50 ml, 19/26 NS
Catalog # WITG2.620.050
Supplier:   witeg Labortechnik
Separating funnels, spherical
Catalog # WITG2.620.050
Supplier:  witeg Labortechnik
Separating funnels, spherical
Separating funnels, spherical
50 ml, 19/26 NS
Price
115,00 €
Log in for availability
1 * 2 ST
Product Details & Documents
Separatory funnel, globe (ball) shaped, made of borosilicate glass 3.3, with detachable PTFE-stopcock and PE stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    50 ml